Assay Details
Target Gene Details
Entrez Gene ID: | 10766 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
APRO5, TOB4, TOBL, TROB2 |
Location: |
Chr.22:41433488-41448230 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOB2 | NM_016272.3 | 2 | 1238 | NP_057356.1 |
XM_005261315.2 | 2 | 2449 | XP_005261372.1 | |
XM_006724105.3 | 2 | 1755 | XP_006724168.1 | |
XM_017028539.1 | 2 | 3531 | XP_016884028.1 | |
AB035207.1 | 1 | 745 | BAA87042.1 | |
AB051450.1 | 2 | 997 | ||
BC028919.1 | 2 | 987 | AAH28919.1 | |
BC038957.1 | 2 | 1011 | AAH38957.1 | |
CR456594.1 | 2 | 691 | CAG30480.1 | |
CR541684.1 | 1 | 531 | CAG46485.1 | |
D64109.1 | 1 | 624 | BAA10971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834207 | Chr.22:41282650 - 41463790 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv589050 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)