Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLC1 | NM_001130107.1 | 16 | 2427 | NP_001123579.1 |
NM_182923.3 | 15 | 2296 | NP_891553.2 | |
AL133587.1 | 2 | 1070 | ||
AY180165.3 | AAO62550.1 | |||
AY180168.1 | 5 | 633 | AAO62553.1 | |
BM988840.1 | 1 | 104 | ||
L04733.1 | 15 | 2252 | AAA16576.1 |
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | ||||
BC001036.1 | AAH01036.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | KLC1 LOC107984648 XRCC3 |
esv2749175 | Chr.14:103700180 - 103705264 on Build GRCh38 | Deletion | KLC1 XRCC3 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | KLC1 ZFYVE21 LOC107984648 XRCC3 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | KLC1 ZFYVE21 LOC107984648 XRCC3 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | KLC1 ZFYVE21 LOC107984648 XRCC3 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | APOPT1 C14orf79 C14orf2 LOC105370708 LINC00638 MIR4710 PPP1R13B BRF1 LOC105370687 JAG2 LOC105370691 PACS2 AHNAK2 NUDT14 KLC1 SIVA1 SNORA28 C14orf180 TMEM179 LINC00637 ASPG LOC102723354 MARK3 MIR203A LOC388022 MIR6765 ZFYVE21 CEP170B PLD4 CKB RD3L LOC107984648 TDRD9 GPR132 EIF5 KIF26A LOC101929634 BTBD6 ZBTB42 LOC105378183 MIR203B BAG5 INF2 XRCC3 ADSSL1 TRMT61A CDCA4 AKT1 |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | KLC1 ZFYVE21 C14orf2 RD3L LINC00637 LOC107984648 PPP1R13B XRCC3 TDRD9 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |