Assay Details
Target Gene Details
Entrez Gene ID: | 5294 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma |
Gene Aliases: |
PI3CG, PI3K, PI3Kgamma, PIK3, p110gamma, p120-PI3K |
Location: |
Chr.7:106865278-106908978 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIK3CG | NM_001282426.1 | 2 | 2010 | NP_001269355.1 |
NM_001282427.1 | 2 | 1934 | NP_001269356.1 | |
NM_002649.3 | 2 | 2159 | NP_002640.2 | |
XM_005250443.3 | 2 | 2027 | XP_005250500.1 | |
XM_011516316.1 | 2 | 1992 | XP_011514618.1 | |
XM_011516317.1 | 2 | 1992 | XP_011514619.1 | |
XM_017012328.1 | 2 | 1992 | XP_016867817.1 | |
AF327656.1 | 1 | 1849 | AAG61115.1 | |
AK292684.1 | 2 | 1934 | ||
AK313753.1 | 2 | 1916 | ||
BC035683.1 | 2 | 1902 | AAH35683.1 | |
BX648341.1 | 2 | 1993 | ||
X83368.1 | 2 | 2172 | CAA58284.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |