Assay Details
Target Gene Details
Entrez Gene ID: | 161931 |
Gene Name: | adenosine deaminase domain containing 2 |
Gene Aliases: |
TENRL |
Location: |
Chr.16:84191117-84197166 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAD2 | NM_001145400.1 | 9 | 1714 | NP_001138872.1 |
NM_139174.3 | 10 | 1960 | NP_631913.3 | |
XM_005255814.1 | 9 | 1744 | XP_005255871.1 | |
XM_011522919.1 | 10 | 1930 | XP_011521221.1 | |
XM_011522920.1 | 10 | 1655 | XP_011521222.1 | |
AF447586.1 | 9 | 1652 | AAM22869.1 | |
AK093046.1 | 10 | 1939 | BAC04032.1 | |
AK093523.1 | 7 | 2214 | ||
AK131102.1 | 9 | 4249 | BAC85152.1 | |
AK315171.1 | 9 | 1633 | ||
BC033491.1 | 9 | 1652 | AAH33491.1 | |
BU731201.1 | 2 | 363 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639413 | Chr.16:84172318 - 84253359 on Build GRCh38 | Loss | KCNG4 TAF1C ADAD2 DNAAF1 |
nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss | KCNG4 MBTPS1 TAF1C LOC729887 ADAD2 DNAAF1 HSDL1 |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss | KCNG4 MBTPS1 SLC38A8 OSGIN1 TAF1C NECAB2 LOC105371371 LOC729887 ADAD2 MLYCD DNAAF1 HSDL1 |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain | KCNG4 MBTPS1 SLC38A8 TAF1C WFDC1 LOC729887 LOC105376777 ADAD2 DNAAF1 HSDL1 |
nsv523437 | Chr.16:84176132 - 84196770 on Build GRCh38 | Loss | TAF1C ADAD2 DNAAF1 |
esv3639414 | Chr.16:84176251 - 84352273 on Build GRCh38 | Loss | KCNG4 TAF1C WFDC1 LOC105376777 ADAD2 DNAAF1 |
nsv457597 | Chr.16:84182086 - 84215151 on Build GRCh38 | Loss | TAF1C ADAD2 |
esv3639412 | Chr.16:84170074 - 84221406 on Build GRCh38 | Loss | KCNG4 TAF1C ADAD2 DNAAF1 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | FLJ30679 KIAA0513 LINC00917 LOC102724344 GINS2 WFDC1 GSE1 MIR1910 ADAD2 ATP2C2 MIR6774 LOC146513 COX4I1 COTL1 TAF1C LOC101928557 CRISPLD2 LOC400548 FENDRR IRF8 KCNG4 FOXF1 MIR5093 KLHL36 EMC8 LOC440390 LOC105371374 LOC105376777 LOC101928614 FOXL1 MTHFSD TLDC1 LINC00311 FAM92B USP10 LOC101928474 ZDHHC7 FOXC2-AS1 LINC01082 FOXC2 LINC01081 DNAAF1 C16orf74 |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss | KCNG4 MBTPS1 SLC38A8 TAF1C LOC729887 ADAD2 DNAAF1 HSDL1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |