Assay Details
Target Gene Details
Entrez Gene ID: | 6476 |
Gene Name: | sucrase-isomaltase |
Gene Aliases: |
- |
Location: |
Chr.3:164978898-165078495 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 35 - Intron 36 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SI | NM_001041.3 | NP_001032.2 | ||
XM_011513078.2 | XP_011511380.1 | |||
BC115034.1 | AAI15035.1 | |||
BC116452.1 | ||||
BC132834.1 | ||||
BC132860.1 | ||||
X63597.1 | CAA45140.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575627 | Chr.3:164978711 - 165081291 on Build GRCh38 | Gain |
![]() |
nsv1014800 | Chr.3:164983237 - 165073353 on Build GRCh38 | Gain |
![]() |
nsv592446 | Chr.3:164547652 - 165237953 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv1013393 | Chr.3:164998562 - 165035342 on Build GRCh38 | Loss |
![]() |
nsv592450 | Chr.3:164988854 - 165078852 on Build GRCh38 | Gain |
![]() |
nsv428426 | Chr.3:164382966 - 165879735 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2763324 | Chr.3:164978854 - 165073365 on Build GRCh38 | Gain |
![]() |
dgv8717n54 | Chr.3:164935636 - 165075970 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)