Assay Details
Target Gene Details
Entrez Gene ID: | 221264 |
Gene Name: | adenylate kinase 9 |
Gene Aliases: |
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1 |
Location: |
Chr.6:109492855-109691253 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AK9 | NM_001145128.2 | 35 | 4850 | NP_001138600.2 |
XM_006715376.3 | 36 | 5715 | XP_006715439.2 | |
XM_011535550.2 | 36 | 5730 | XP_011533852.1 | |
XM_011535552.2 | 35 | 4906 | XP_011533854.1 | |
XM_011535553.2 | 35 | 5513 | XP_011533855.1 | |
XM_011535554.2 | 34 | 5409 | XP_011533856.1 | |
XM_011535558.1 | 17 | 2751 | XP_011533860.1 | |
XM_017010384.1 | 36 | 5730 | XP_016865873.1 | |
XM_017010385.1 | 35 | 4898 | XP_016865874.1 | |
XM_017010386.1 | 36 | 5730 | XP_016865875.1 | |
XM_017010387.1 | 30 | 5361 | XP_016865876.1 | |
XM_017010389.1 | 21 | 3613 | XP_016865878.1 | |
AK098657.1 | 1 | 88 | BAC05368.1 | |
BC101714.1 | AAI01715.1 | |||
BC111948.1 | AAI11949.1 | |||
BC143533.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115666311] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |