Assay Details
Target Gene Details
Entrez Gene ID: | 8464 |
Gene Name: | SPT3 homolog, SAGA and STAGA complex component |
Gene Aliases: |
SPT3, SPT3L |
Location: |
Chr.6:44808844-45378051 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUPT3H | NM_001261823.1 | 10 | 1963 | NP_001248752.1 |
NM_003599.3 | 11 | 2054 | NP_003590.1 | |
NM_181356.2 | 13 | 2239 | NP_852001.1 | |
XM_011514949.2 | 14 | 2146 | XP_011513251.1 | |
XM_011514950.1 | 12 | 2125 | XP_011513252.1 | |
XM_011514951.2 | 12 | 6186 | XP_011513253.1 | |
XM_011514952.2 | 12 | 3172 | XP_011513254.1 | |
XM_011514953.2 | 12 | 1955 | XP_011513255.1 | |
XM_011514954.2 | 12 | 1986 | XP_011513256.1 | |
XM_017011369.1 | 14 | 6374 | XP_016866858.1 | |
XM_017011370.1 | 12 | 1939 | XP_016866859.1 | |
XM_017011371.1 | 12 | 5229 | XP_016866860.1 | |
XM_017011372.1 | 11 | 1943 | XP_016866861.1 | |
XM_017011373.1 | 12 | 1996 | XP_016866862.1 | |
XM_017011374.1 | 11 | 1821 | XP_016866863.1 | |
AK092340.1 | 1 | 322 | ||
BC035816.1 | 11 | 1931 | AAH35816.1 | |
BC050384.1 | 13 | 2121 | AAH50384.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |