Assay Details
Target Gene Details
Entrez Gene ID: | 1609 |
Gene Name: | diacylglycerol kinase theta |
Gene Aliases: |
DAGK, DAGK4, DAGK7 |
Location: |
Chr.4:958884-973560 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DGKQ | NM_001347.3 | 22 | 2730 | NP_001338.2 |
XM_011513411.1 | 22 | 2715 | XP_011511713.1 | |
AB209503.1 | 22 | 2481 | BAD92740.1 | |
BC063801.1 | 22 | 2738 | AAH63801.1 | |
GU727647.1 | 22 | 2668 | ||
L38707.1 | 22 | 2690 | AAA98749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003219 | Chr.4:776106 - 1111676 on Build GRCh38 | Gain | CPLX1 DGKQ GAK RNF212 TMEM175 SLC26A1 LOC100129917 IDUA FGFRL1 |
nsv593226 | Chr.4:948218 - 1025223 on Build GRCh38 | Loss | DGKQ TMEM175 SLC26A1 IDUA FGFRL1 |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | SPON2 PDE6B RNF212 CTBP1-AS TMEM175 SLC26A1 LOC105374344 CRIPAK LOC101928548 LOC101928521 ATP5I TMED11P CPLX1 CTBP1-AS2 LOC100130872 LOC107986211 LOC100129917 CTBP1 MFSD7 MYL5 IDUA FGFRL1 UVSSA PCGF3 DGKQ GAK MAEA NKX1-1 |
nsv950327 | Chr.4:945913 - 978512 on Build GRCh38 | Deletion | DGKQ TMEM175 |
dgv5079n100 | Chr.4:843485 - 970603 on Build GRCh38 | Gain | DGKQ GAK TMEM175 |
esv29984 | Chr.4:857569 - 970571 on Build GRCh38 | Loss | DGKQ GAK TMEM175 |
nsv469998 | Chr.4:815805 - 1093743 on Build GRCh38 | Loss | CPLX1 DGKQ GAK RNF212 TMEM175 SLC26A1 IDUA FGFRL1 |
nsv1008352 | Chr.4:819605 - 1041628 on Build GRCh38 | Gain | CPLX1 DGKQ GAK TMEM175 SLC26A1 IDUA FGFRL1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |