Assay Details
Target Gene Details
Entrez Gene ID: | 56910 |
Gene Name: | StAR related lipid transfer domain containing 7 |
Gene Aliases: |
GTT1 |
Location: |
Chr.2:96184859-96208846 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7 | NM_020151.3 | 8 | 3035 | NP_064536.2 |
XM_011511499.1 | 8 | 2652 | XP_011509801.1 | |
AB075479.1 | 1 | 661 | ||
AF270647.1 | 8 | 2962 | AAF81750.1 | |
AK023234.1 | 7 | 2294 | ||
AK054615.1 | 8 | 2401 | ||
AK094925.1 | 2 | 2607 | ||
AK096666.1 | 5 | 1795 | ||
BC007894.2 | 9 | 2793 | AAH07894.1 | |
BC008894.2 | 8 | 3018 | AAH08894.1 | |
BC009998.2 | 9 | 2793 | AAH09998.1 | |
BC012774.2 | 8 | 2985 | AAH12774.1 | |
BC012793.2 | 8 | 2999 | AAH12793.1 | |
BC013279.2 | 8 | 3018 | AAH13279.1 | |
BC014076.2 | 8 | 2995 | AAH14076.1 | |
BC014274.2 | 8 | 3014 | AAH14274.3 | |
BC032106.1 | 8 | 3009 | AAH32106.1 | |
CR749695.1 | 1 | 1350 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain | STARD7-AS1 NCAPH LOC105373495 STARD7 SNRNP200 TMEM127 CIAO1 ITPRIPL1 |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | MIR3127 LOC101927053 LOC105373495 ANKRD36B LOC107984110 KANSL3 ANKRD39 LOC100506076 FAHD2CP STARD7-AS1 NCAPH NEURL3 CNNM3 SNRNP200 DUSP2 ARID5A ANKRD36C ITPRIPL1 FAHD2B ADRA2B FAM178B CNNM4 LMAN2L STARD7 ANKRD36 ANKRD23 LOC653924 TMEM127 LOC100506123 ASTL CIAO1 LOC105373496 GPAT2 SEMA4C FER1L5 |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | MIR3127 LOC101927053 LOC105373495 ANKRD36B LOC107984110 KANSL3 ANKRD39 LOC100506076 FAHD2CP STARD7-AS1 NCAPH NEURL3 CNNM3 SNRNP200 DUSP2 ARID5A ANKRD36C ITPRIPL1 FAHD2B ADRA2B FAM178B CNNM4 LMAN2L STARD7 ANKRD36 ANKRD23 LOC653924 TMEM127 LOC100506123 ASTL CIAO1 LOC105373496 GPAT2 SEMA4C FER1L5 |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | MIR3127 LOC101927053 LOC105373495 ANKRD36B LOC107984110 KANSL3 ANKRD39 LOC100506076 FAHD2CP STARD7-AS1 NCAPH NEURL3 CNNM3 SNRNP200 DUSP2 ARID5A ANKRD36C ITPRIPL1 FAHD2B ADRA2B FAM178B CNNM4 LMAN2L STARD7 ANKRD36 ANKRD23 LOC653924 TMEM127 LOC100506123 ASTL CIAO1 LOC105373496 GPAT2 SEMA4C FER1L5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |