Assay Details
Target Gene Details
Entrez Gene ID: | 85441 |
Gene Name: | helicase with zinc finger 2 |
Gene Aliases: |
PDIP-1, PRIC285 |
Location: |
Chr.20:63558086-63574239 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HELZ2 | NM_001037335.2 | 20 | 8792 | NP_001032412.2 |
NM_033405.3 | 14 | 6297 | NP_208384.3 | |
AB051556.2 | 14 | 6297 | ||
AB201715.1 | 19 | 7970 | BAE46995.1 | |
AF517673.1 | 14 | 6617 | AAM74197.1 | |
AK055611.1 | 9 | 1329 | BAB70969.1 | |
AK074171.1 | 12 | 3326 | BAB84997.1 | |
BC150283.1 | 14 | 6297 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv586578 | Chr.20:63558992 - 63570431 on Build GRCh38 | Loss |
![]() |
nsv953306 | Chr.20:63151949 - 63571247 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv586573 | Chr.20:63535645 - 63560584 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv521734 | Chr.20:63522433 - 63566995 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2722937 | Chr.20:63559306 - 63559887 on Build GRCh38 | Deletion |
![]() |
nsv470563 | Chr.20:63486715 - 63793308 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834034 | Chr.20:63462825 - 63625901 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv517702 | Chr.20:63540916 - 63581575 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv2758806 | Chr.20:63470996 - 63745014 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)