Assay Details
Target Gene Details
Entrez Gene ID: | 25925 |
Gene Name: | zinc finger protein 521 |
Gene Aliases: |
EHZF, Evi3 |
Location: |
Chr.18:25061924-25352250 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF521 | NM_001308225.1 | 6 | 4067 | NP_001295154.1 |
NM_015461.2 | 7 | 4108 | NP_056276.1 | |
XM_011525909.2 | 7 | 4105 | XP_011524211.1 | |
XM_011525911.1 | 8 | 4144 | XP_011524213.1 | |
XM_017025697.1 | 6 | 3994 | XP_016881186.1 | |
XM_017025698.1 | 8 | 4071 | XP_016881187.1 | |
AF141339.1 | 4 | 522 | AAG49442.1 | |
AJ518106.1 | 7 | 4005 | CAD57322.1 | |
AK021452.1 | 4 | 2172 | BAB13829.1 | |
AK027354.1 | 7 | 4010 | BAB55056.1 | |
AK074046.1 | 4 | 3630 | ||
AK293067.1 | 7 | 4059 | ||
AL117615.1 | 4 | 1677 | CAB56016.1 | |
BC032869.2 | 6 | 3966 | AAH32869.2 | |
BC113622.1 | 7 | 3929 | ||
BC113648.1 | 7 | 3929 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |