Assay Details
Target Gene Details
Entrez Gene ID: | 81889 |
Gene Name: | fumarylacetoacetate hydrolase domain containing 1 |
Gene Aliases: |
C16orf36, YISKL |
Location: |
Chr.16:1827224-1840207 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAHD1 | NM_001018104.2 | 1 | 597 | NP_001018114.1 |
NM_001142398.1 | 1 | 597 | NP_001135870.1 | |
NM_031208.3 | 1 | 597 | NP_112485.1 | |
AK094199.1 | 1 | 596 | BAC04308.1 | |
AL136720.1 | 1 | 597 | CAB66654.1 | |
BC063017.1 | 1 | 601 | AAH63017.1 | |
CR533499.1 | 1 | 591 | CAG38530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517196 | Chr.16:1729694 - 1844960 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv952900 | Chr.16:1724500 - 1827999 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)