Assay Details
Target Gene Details
Entrez Gene ID: | 10930 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 2 |
Gene Aliases: |
ARCD1, ARP1 |
Location: |
Chr.6:41053201-41064891 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOBEC2 | NM_006789.3 | 3 | 1119 | NP_006780.1 |
AF086496.1 | ||||
AF161698.1 | 3 | 1027 | AAD45360.1 | |
AI096570.1 | 1 | 142 | ||
AK223461.1 | BAD97181.1 | |||
AK300689.1 | 4 | 893 | ||
AK308197.1 | 3 | 950 | ||
AK310006.1 | 5 | 931 | ||
BC047767.2 | 3 | 1056 | AAH47767.1 | |
BC069688.1 | 3 | 1016 | AAH69688.1 | |
BC069764.1 | 3 | 1019 | AAH69764.1 | |
CB051273.1 | 1 | 166 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |