Assay Details
Target Gene Details
Entrez Gene ID: | 4688 |
Gene Name: | neutrophil cytosolic factor 2 |
Gene Aliases: |
NCF-2, NOXA2, P67-PHOX, P67PHOX |
Location: |
Chr.1:183555562-183590921 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCF2 | NM_000433.3 | 15 | 2303 | NP_000424.2 |
NM_001127651.2 | 16 | 2140 | NP_001121123.1 | |
NM_001190789.1 | 13 | 2060 | NP_001177718.1 | |
NM_001190794.1 | 14 | 2168 | NP_001177723.1 | |
XM_005245207.1 | 14 | 2124 | XP_005245264.1 | |
XM_011509580.1 | 16 | 2141 | XP_011507882.1 | |
XM_011509581.1 | 16 | 2128 | XP_011507883.1 | |
AB209647.1 | 13 | 5573 | BAD92884.1 | |
AW951155.1 | 1 | 466 | ||
BM979513.1 | 1 | 127 | ||
BQ029626.1 | 1 | 126 | ||
BU617086.1 | 1 | 127 | ||
M32011.1 | 16 | 2095 | AAA36379.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |