Assay Details
Target Gene Details
Entrez Gene ID: | 9354 |
Gene Name: | ubiquitination factor E4A |
Gene Aliases: |
E4, UBOX2, UFD2 |
Location: |
Chr.11:118359581-118399211 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE4A | NM_001204077.1 | 5 | 579 | NP_001191006.1 |
NM_004788.3 | 5 | 579 | NP_004779.2 | |
AF087968.1 | ||||
AK296369.1 | 5 | 526 | ||
BC030973.1 | 5 | 528 | ||
BC052643.1 | 5 | 537 | AAH52643.1 | |
BC062695.1 | 5 | 559 | AAH62695.1 | |
BC111417.1 | 5 | 553 | ||
BC112367.1 | 5 | 556 | AAI12368.1 | |
BU167020.1 | 5 | 536 | ||
D50916.1 | 5 | 516 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |