Assay Details
Target Gene Details
Entrez Gene ID: | 84690 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
NYD-SP20, NYDSP20 |
Location: |
Chr.17:3440011-3513852 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA22 | NM_001170695.1 | 8 | 1035 | NP_001164166.1 |
NM_001170696.1 | 7 | 906 | NP_001164167.1 | |
NM_001170697.1 | 8 | 1041 | NP_001164168.1 | |
NM_001170698.1 | 8 | 1056 | NP_001164169.1 | |
NM_001170699.1 | NP_001164170.1 | |||
NM_001321336.1 | 8 | 1325 | NP_001308265.1 | |
NM_001321337.1 | 8 | 1373 | NP_001308266.1 | |
NM_032598.4 | 8 | 1097 | NP_115987.2 | |
AF367472.1 | 8 | 989 | AAK53408.1 | |
AK295299.1 | 8 | 1325 | ||
AK301892.1 | ||||
AY032684.1 | 7 | 855 | AAK51120.1 | |
AY035867.1 | 8 | 1038 | AAK61373.1 | |
AY035868.1 | 8 | 1097 | AAK61374.1 | |
BC029483.1 | 8 | 982 | AAH29483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892950 | Chr.17:3131812 - 3577153 on Build GRCh38 | Loss | OR3A1 TRPV1 OR1E1 OR1A2 OR3A3 TRPV3 OR1A1 SPATA22 OR1E2 OR3A4P OR1D4 OR3A2 ASPA |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain | CAMKK1 OR1G1 CCDC92B LOC105371592 OR1E1 ITGAE CTNS OR1A2 P2RX1 TRPV3 OR1A1 SHPK EMC6 MIR1253 OR1D5 RAP1GAP2 OR1E2 OR3A4P LOC105371591 CLUH P2RX5 GSG2 P2RX5-TAX1BP3 OR3A1 PAFAH1B1 TRPV1 LOC101927911 ATP2A3 OR3A3 MIR6776 SPATA22 NCBP3 TAX1BP3 OR1D2 OR1D4 OR3A2 ASPA |
nsv574232 | Chr.17:3254715 - 3503170 on Build GRCh38 | Gain | OR1E2 OR3A1 OR3A4P OR1E1 OR3A3 OR3A2 SPATA22 ASPA |
esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion | OR1G1 CCDC92B OR3A1 PAFAH1B1 LOC105371592 LOC101927911 OR1E1 OR1A2 OR3A3 TRPV3 OR1A1 MIR6776 SPATA22 MIR1253 OR1D5 OR1D2 RAP1GAP2 OR1E2 OR3A4P LOC105371591 CLUH OR1D4 OR3A2 ASPA |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain | CAMKK1 GSG2 P2RX5-TAX1BP3 TRPV1 OR1E1 ITGAE CTNS OR3A3 TRPV3 SPATA22 NCBP3 SHPK EMC6 TAX1BP3 OR1E2 OR3A2 P2RX5 ASPA |
nsv521277 | Chr.17:3390044 - 3468885 on Build GRCh38 | Gain | OR1E2 OR1E1 OR3A3 SPATA22 |
nsv833341 | Chr.17:3352243 - 3521390 on Build GRCh38 | Loss | OR1E2 OR1E1 OR3A3 OR3A2 TRPV3 SPATA22 ASPA |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss | OR1E2 TRPV1 OR3A3 TRPV3 SPATA22 SHPK ASPA |
nsv1059413 | Chr.17:3365789 - 3453922 on Build GRCh38 | Loss | OR1E2 OR1E1 OR3A3 OR3A2 SPATA22 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |