Assay Details
Target Gene Details
Entrez Gene ID: | 57010 |
Gene Name: | calcium binding protein 4 |
Gene Aliases: |
CRSD, CSNB2B |
Location: |
Chr.11:67452406-67461774 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CABP4 | NM_001300895.1 | NP_001287824.1 | ||
NM_001300896.1 | NP_001287825.1 | |||
NM_145200.3 | NP_660201.1 | |||
XM_005274114.3 | XP_005274171.2 | |||
XM_011545181.2 | XP_011543483.1 | |||
XM_011545182.2 | XP_011543484.1 | |||
XM_011545183.2 | XP_011543485.1 | |||
XM_017018025.1 | XP_016873514.1 | |||
AL712439.1 | ||||
AY039217.1 | AAK83462.1 | |||
BC033167.1 | AAH33167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | RPS6KB2 GPR152 CARNS1 CABP4 CORO1B CDK2AP2 AIP PTPRCAP PITPNM1 TMEM134 MIR6752 |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | RPS6KB2 GPR152 CABP4 TBC1D10C CABP2 AIP PTPRCAP RAD9A PITPNM1 TMEM134 CARNS1 CORO1B CDK2AP2 PPP1CA MIR6752 |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | LOC100130987 RPS6KB2 GPR152 CABP4 TBC1D10C CABP2 AIP PTPRCAP RAD9A PITPNM1 TMEM134 CLCF1 CARNS1 CORO1B CDK2AP2 PPP1CA MIR6752 |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | RPS6KB2 GPR152 CABP4 TBC1D10C CABP2 AIP PTPRCAP PITPNM1 TMEM134 CARNS1 CORO1B CDK2AP2 MIR6752 |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | GPR152 CABP4 CORO1B CDK2AP2 CABP2 AIP PITPNM1 TMEM134 MIR6752 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |