Assay Details
Target Gene Details
Entrez Gene ID: | 23341 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C16 |
Gene Aliases: |
- |
Location: |
Chr.1:15526694-15571733 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNAJC16 | NM_001287811.1 | 3 | 469 | NP_001274740.1 |
NM_015291.3 | 4 | 654 | NP_056106.1 | |
NR_109898.1 | 4 | 654 | ||
AB023179.2 | 4 | 610 | ||
AK091848.1 | 4 | 612 | ||
AK292479.1 | 4 | 586 | ||
AL833299.1 | 4 | 593 | ||
BC047363.1 | 4 | 654 | AAH47363.1 | |
BX648982.1 | 4 | 579 | ||
CR749259.1 | 3 | 414 | CAH18115.1 | |
CR749567.1 | 3 | 414 | CAH18363.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |