Assay Details
Target Gene Details
Entrez Gene ID: | 79568 |
Gene Name: | chromosome 2 open reading frame 47 |
Gene Aliases: |
- |
Location: |
Chr.2:199955317-199964125 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C2orf47 | NM_024520.2 | 5 | 1051 | NP_078796.2 |
XM_005246852.2 | XP_005246909.1 | |||
AK026208.1 | 5 | 1051 | BAB15393.1 | |
AL832964.1 | 4 | 370 | CAH56348.1 | |
BC017959.1 | 4 | 735 | AAH17959.1 | |
BQ448209.1 | 2 | 475 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834510 | Chr.2:199852719 - 200052794 on Build GRCh38 | Loss | C2orf69 TYW5 C2orf47 |