Assay Details
Target Gene Details
Entrez Gene ID: | 51005 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
CGI-14 |
Location: |
Chr.16:2520351-2531414 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMDHD2 | NM_001145815.1 | 5 | 596 | NP_001139287.1 |
NM_015944.3 | 5 | 596 | NP_057028.2 | |
XM_017023262.1 | 5 | 607 | XP_016878751.1 | |
XM_017023263.1 | 5 | 607 | XP_016878752.1 | |
XM_017023264.1 | 5 | 607 | XP_016878753.1 | |
XM_017023265.1 | 5 | 607 | XP_016878754.1 | |
XM_017023266.1 | XP_016878755.1 | |||
XM_017023267.1 | 4 | 459 | XP_016878756.1 | |
AB209351.1 | 2 | 3870 | BAD92588.1 | |
AF132948.1 | 5 | 586 | AAD27723.1 | |
AK296877.1 | 5 | 568 | ||
AK300747.1 | 4 | 412 | ||
BC018734.1 | 5 | 532 | AAH18734.1 | |
CN362870.1 | 4 | 466 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | NHLRC4 MSRB1 SPSB3 KCTD5 WFIKKN1 GNPTG GNG13 MIR3178 TPSAB1 METTL26 SSTR5-AS1 LOC105371047 CASKIN1 STUB1 UBE2I PGP SNORA78 LUC7L MIR4516 WDR90 HAGH RNF151 SLC9A3R2 LOC101929280 HN1L MIR940 C16orf59 LOC107984876 TELO2 SNORD60 CRAMP1 RAB26 RPUSD1 PRR25 TPSB2 MIR6768 CHTF18 AMDHD2 PIGQ MIR3176 NME4 HBM CCDC154 LOC105371049 NOXO1 PKD1 ARHGDIG C16orf91 ERVK13-1 DDX11L10 LINC00235 NPRL3 LOC105371038 LOC100134368 PDPK1 LOC729652 IGFALS MIR3677 RHBDL1 FAM234A TMEM204 BAIAP3 HBQ1 HS3ST6 MEIOB RPL23AP5 TPSD1 MRPS34 SNHG9 SOX8 NDUFB10 PTX4 LOC105371045 MIR662 RHBDF1 NPW HAGHL TMEM8A CLCN7 TBL3 TSR3 NTHL1 FLJ42627 WASIR2 PRSS27 IFT140 LOC105371184 WDR24 HBA1 LOC100287175 LOC107987233 FBXL16 ABCA3 NARFL MAPK8IP3 SNORA10 CCDC78 NUBP2 RPL3L C1QTNF8 LINC00254 MIR6511B1 SSTR5 CAPN15 LOC652276 TSC2 MIR3177 LMF1-AS1 MPG E4F1 ATP6V0C ECI1 EME2 TRAF7 TBC1D24 MIR6859-4 ABCA17P SNRNP25 LMF1 TPSG1 POLR3K MRPL28 SYNGR3 HBZ MIR3180-5 RHOT2 METRN CACNA1H MSLN RPS2 MIR5587 CEMP1 DNASE1L2 RNPS1 NME3 PRSS29P LOC105371046 RAB40C NTN3 MIR6767 LOC106660606 HBA2 PRR35 FAM173A DECR2 ZNF598 GFER MIR4717 BRICD5 UNKL MCRIP2 RAB11FIP3 CCNF JMJD8 PDIA2 RGS11 FAHD1 MLST8 SNORA64 AXIN1 SNHG19 MIR1225 |
esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | PRSS27 SRRM2 TCEB2 PRSS33 SRRM2-AS1 LOC652276 AMDHD2 KCTD5 CEMP1 MIR3178 PRSS41 ATP6V0C ERVK13-1 TBC1D24 PDPK1 FLJ42627 PRSS21 |
nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss | ATP6V0C ERVK13-1 LOC652276 TBC1D24 PDPK1 AMDHD2 CEMP1 FLJ42627 MIR3178 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | MIR3180-5 MIR6768 AMDHD2 CEMP1 DNASE1L2 RNPS1 ABCA3 MIR3178 LOC105371049 PKD1 NTN3 CASKIN1 PDPK1 LOC729652 MIR6767 MIR3677 LOC106660606 MIR6511B1 PGP LOC652276 TSC2 MIR4516 MIR4717 BRICD5 E4F1 ATP6V0C MIR940 C16orf59 ECI1 CCNF TRAF7 TBC1D24 ABCA17P SNORD60 MLST8 SNHG19 MIR1225 RAB26 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |