Assay Details
Target Gene Details
Entrez Gene ID: | 8848 |
Gene Name: | TSC22 domain family member 1 |
Gene Aliases: |
Ptg-2, TGFB1I4, TSC22 |
Location: |
Chr.13:44432143-44576565 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSC22D1 | NM_001243797.1 | 3 | 195 | NP_001230726.1 |
NM_001243798.1 | 3 | 200 | NP_001230727.1 | |
NM_001243799.1 | NP_001230728.1 | |||
NM_006022.3 | 3 | 537 | NP_006013.1 | |
NM_183422.3 | 3 | 3519 | NP_904358.2 | |
AB082525.1 | 3 | 3519 | ||
AJ222700.1 | 3 | 432 | CAA10951.1 | |
AK027071.1 | 3 | 2074 | ||
AK091854.1 | ||||
AK290751.1 | 3 | 472 | ||
AK312345.1 | 3 | 471 | ||
BC000456.2 | 3 | 453 | AAH00456.1 | |
BC016867.1 | 3 | 456 | AAH16867.1 | |
BC069207.1 | 3 | 445 | AAH69207.1 | |
BC105277.1 | 3 | 448 | AAI05278.1 | |
BC136475.1 | 3 | 3289 | ||
BC136482.1 | 3 | 3289 | ||
BC146664.1 | 3 | 452 | ||
BC146679.1 | 3 | 452 | ||
BF307401.1 | 3 | 197 | ||
BG703493.1 | 3 | 543 | ||
BI463370.1 | 3 | 203 | ||
CN344671.1 | 3 | 175 | ||
CR456711.1 | 3 | 241 | CAG32992.1 | |
CR627459.1 | 4 | 1712 | CAH10539.1 | |
D38585.1 | 3 | 241 | BAA07598.1 | |
D87061.1 | 3 | 241 | BAB46917.1 | |
U35048.1 | 3 | 423 | AAC50566.1 | |
X97299.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |