Assay Details
Target Gene Details
Entrez Gene ID: | 9146 |
Gene Name: | hepatocyte growth factor-regulated tyrosine kinase substrate |
Gene Aliases: |
HRS |
Location: |
Chr.17:81683932-81702121 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HGS | NM_004712.4 | NP_004703.1 | ||
XM_011525463.2 | XP_011523765.1 | |||
XM_017025297.1 | XP_016880786.1 | |||
XM_017025298.1 | XP_016880787.1 | |||
AF260566.1 | AAF82361.1 | |||
AK294190.1 | ||||
AK303800.1 | ||||
BC003565.1 | AAH03565.1 | |||
BT009754.1 | AAP88756.1 | |||
D84064.1 | BAA23366.1 | |||
DR001213.1 | ||||
KU178466.1 | ||||
KU178467.1 | ||||
U43895.1 | AAC51929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC107985021 NPLOC4 LOC107985022 ARL16 MIR4740 MIR338 OXLD1 CCDC137 BAIAP2 ACTG1 MIR657 MIR6786 SLC38A10 MIR1250 LOC400627 MRPL12 PDE6G FSCN2 TEPSIN TSPAN10 BAHCC1 BAIAP2-AS1 AATK-AS1 LOC105371925 AATK LOC105371921 LOC100129503 RPTOR HGS LOC101928855 LOC100130370 SLC25A10 CHMP6 FAAP100 C17orf89 MIR3065 LINC00482 CEP131 MIR3186 TMEM105 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | MIR6787 OGFOD3 LOC105376790 ARL16 LOC101929552 MAFG LOC105371944 NARF CCDC137 NOTUM SLC38A10 DUS1L GPS1 MCRIP1 SLC16A3 PYCR1 CSNK1D MIR4525 MRPL12 LOC105376791 FSCN2 MAFG-AS1 WDR45B C17orf62 BAHCC1 AATK RAC3 FASN PCYT2 ARHGDIA FN3KRP RFNG ASPSCR1 SLC25A10 RAB40B ZNF750 TMEM105 NPLOC4 MIR4740 PPP1R27 OXLD1 CCDC57 LOC101929511 DCXR ACTG1 MIR6786 STRA13 TEX19 SIRT7 PDE6G TEPSIN TSPAN10 SECTM1 NPB ANAPC11 AATK-AS1 LOC105371925 TBCD FOXK2 UTS2R HEXDC HGS GCGR LOC100130370 FAAP100 C17orf89 B3GNTL1 FN3K P4HB MYADML2 SNORD134 LINC00482 LRRC45 CEP131 MIR3186 ALYREF CD7 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | NPLOC4 ARL16 PPP1R27 OXLD1 MAFG CCDC137 DCXR NOTUM MIR6786 STRA13 DUS1L GPS1 MCRIP1 PYCR1 SIRT7 MRPL12 PDE6G FSCN2 MAFG-AS1 TSPAN10 NPB ANAPC11 RAC3 HGS GCGR PCYT2 ARHGDIA RFNG ASPSCR1 SLC25A10 FAAP100 P4HB MYADML2 LRRC45 ALYREF |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | ARL16 PPP1R27 OXLD1 CCDC137 HGS GCGR MIR6786 SLC25A10 MCRIP1 P4HB MRPL12 PDE6G TSPAN10 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |