Assay Details
Target Gene Details
Entrez Gene ID: | 51272 |
Gene Name: | Bet1 golgi vesicular membrane trafficking protein like |
Gene Aliases: |
BET1L1, GOLIM3, GS15, HSPC197 |
Location: |
Chr.11:202924-207422 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BET1L | NM_001098787.1 | 4 | 1754 | NP_001092257.1 |
NM_016526.4 | 3 | 1895 | NP_057610.2 | |
AF151031.1 | 1 | 665 | AAF36117.1 | |
AK022958.1 | 4 | 1742 | ||
AK314273.1 | 1 | 734 | ||
AL833842.1 | 1 | 325 | ||
AY007148.1 | AAG02004.1 | |||
BC032779.2 | 3 | 1855 | AAH32779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625061 | Chr.11:195766 - 233956 on Build GRCh38 | Gain | MIR6743 ODF3 BET1L RIC8A SIRT3 |
esv3625060 | Chr.11:194412 - 252166 on Build GRCh38 | Gain | MIR6743 SCGB1C1 ODF3 BET1L RIC8A SIRT3 PSMD13 |
nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain | MIR6743 IFITM3 IFITM5 ODF3 PGGHG BET1L IFITM1 IFITM2 RIC8A SIRT3 PSMD13 NLRP6 |
esv3625062 | Chr.11:198524 - 203951 on Build GRCh38 | Loss | ODF3 BET1L |
dgv1011n100 | Chr.11:198510 - 275185 on Build GRCh38 | Gain | MIR6743 ODF3 BET1L RIC8A SIRT3 PSMD13 |
esv22231 | Chr.11:60059 - 216243 on Build GRCh38 | Gain | MIR6743 SCGB1C1 ODF3 LOC388572 BET1L RIC8A SIRT3 LINC01001 |
nsv951267 | Chr.11:196001 - 206600 on Build GRCh38 | Deletion | ODF3 BET1L |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |