Assay Details
Target Gene Details
Entrez Gene ID: | 80832 |
Gene Name: | apolipoprotein L4 |
Gene Aliases: |
APOL-IV, APOLIV |
Location: |
Chr.22:36189128-36204833 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOL4 | NM_030643.4 | 6 | 1173 | NP_085146.2 |
NM_145660.2 | 5 | 984 | NP_663693.1 | |
AF305226.1 | 4 | 825 | AAK20212.1 | |
AK225696.1 | 4 | 834 | ||
AK290466.1 | 5 | 1158 | ||
AK290777.1 | 4 | 805 | ||
AK311506.1 | 5 | 957 | ||
AK312716.1 | 4 | 883 | ||
AK313637.1 | 5 | 921 | ||
AY014914.1 | 6 | 1174 | AAG50352.1 | |
AY014915.1 | 5 | 984 | AAG50353.1 | |
CR456380.1 | 4 | 763 | CAG30266.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |