Assay Details
Target Gene Details
Entrez Gene ID: | 113 |
Gene Name: | adenylate cyclase 7 |
Gene Aliases: |
AC7 |
Location: |
Chr.16:50246158-50318135 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADCY7 | NM_001114.4 | 25 | 5656 | NP_001105.1 |
XM_011522835.2 | 26 | 5775 | XP_011521137.1 | |
XM_011522836.2 | 26 | 5791 | XP_011521138.1 | |
XM_011522837.2 | 26 | 5720 | XP_011521139.1 | |
XM_011522838.2 | 26 | 5924 | XP_011521140.1 | |
XM_011522839.2 | 26 | 5751 | XP_011521141.1 | |
XM_011522840.2 | 26 | 5758 | XP_011521142.1 | |
XM_011522842.2 | 26 | 5821 | XP_011521144.1 | |
XM_017022895.1 | 26 | 5773 | XP_016878384.1 | |
XM_017022896.1 | 23 | 4937 | XP_016878385.1 | |
XM_017022897.1 | 23 | 4956 | XP_016878386.1 | |
AA740624.1 | ||||
BM543987.1 | 1 | 251 | ||
BU631614.1 | 1 | 501 | ||
D25538.1 | 25 | 5653 |
Target Gene Details
Entrez Gene ID: | 29117 |
Gene Name: | bromodomain containing 7 |
Gene Aliases: |
BP75, CELTIX1, NAG4 |
Location: |
Chr.16:50317563-50368969 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BRD7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1805 | Chr.16:50307115 - 50352023 on Build GRCh38 | Deletion | ADCY7 BRD7 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |