Assay Details
Target Gene Details
Entrez Gene ID: | 64579 |
Gene Name: | N-deacetylase and N-sulfotransferase 4 |
Gene Aliases: |
N-HSST, N-HSST 4, NDST-4, NHSST4 |
Location: |
Chr.4:114824122-115114098 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDST4 | NM_022569.2 | 11 | 2876 | NP_072091.1 |
XM_017008545.1 | 10 | 1874 | XP_016864034.1 | |
XM_017008546.1 | 9 | 1180 | XP_016864035.1 | |
AB036429.1 | 11 | 2876 | BAB18535.1 | |
AK289670.1 | 11 | 2854 | ||
BC113075.2 | 10 | 1563 | AAI13076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830040 | Chr.4:114814399 - 114986969 on Build GRCh38 | Gain | NDST4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |