Assay Details
Target Gene Details
Entrez Gene ID: | 197021 |
Gene Name: | lactase like |
Gene Aliases: |
KLG, KLPH |
Location: |
Chr.15:66547468-66566025 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LCTL | NM_001278562.1 | 12 | 1622 | NP_001265491.1 |
NM_207338.3 | 13 | 1913 | NP_997221.2 | |
NR_103739.1 | 9 | 1371 | ||
AK090598.1 | 12 | 1607 | ||
AK311441.1 | 9 | 1371 | ||
AY358729.1 | 13 | 1913 | AAQ89091.1 | |
EB361548.1 |
Target Gene Details
Entrez Gene ID: | 55055 |
Gene Name: | zwilch kinetochore protein |
Gene Aliases: |
KNTC1AP, hZwilch |
Location: |
Chr.15:66505083-66549485 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZWILCH | NM_001287821.1 | NP_001274750.1 | ||
NM_001287822.1 | NP_001274751.1 | |||
NM_001287823.1 | NP_001274752.1 | |||
NM_017975.4 | NP_060445.3 | |||
AK023175.1 | BAB14446.1 | |||
AK027468.1 | BAB55133.1 | |||
AK291048.1 | ||||
BC036900.1 | AAH36900.1 | |||
BC090041.1 | AAH90041.1 | |||
BX640701.1 | CAE45821.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |