Assay Details
Target Gene Details
Entrez Gene ID: | 7264 |
Gene Name: | tissue specific transplantation antigen P35B |
Gene Aliases: |
FX, P35B, SDR4E1 |
Location: |
Chr.8:143612618-143618043 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSTA3 | NM_001317783.1 | NP_001304712.1 | ||
NM_003313.3 | NP_003304.1 | |||
XM_005251051.3 | XP_005251108.2 | |||
XM_011517269.1 | XP_011515571.1 | |||
AK096752.1 | 1 | 728 | ||
AK303125.1 | ||||
AK313560.1 | ||||
BC001941.1 | AAH01941.1 | |||
BC093061.1 | AAH93061.1 | |||
BG709420.1 | ||||
CB132195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2118e212 | Chr.8:143616751 - 143643564 on Build GRCh38 | Loss |
![]() ![]() |
nsv1161810 | Chr.8:143542268 - 143622987 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv612793 | Chr.8:143552167 - 143630578 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv12427n54 | Chr.8:143615483 - 143632493 on Build GRCh38 | Loss |
![]() |
nsv612777 | Chr.8:143538914 - 143632493 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv12428n54 | Chr.8:143616727 - 143626978 on Build GRCh38 | Loss |
![]() |
esv3584899 | Chr.8:143611680 - 143636819 on Build GRCh38 | Loss |
![]() ![]() |
nsv42 | Chr.8:143610600 - 143635500 on Build GRCh38 | Deletion |
![]() |
nsv6439 | Chr.8:143606519 - 143651282 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
dgv7336n100 | Chr.8:143510004 - 143616975 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv271n111 | Chr.8:143601764 - 143642893 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv831488 | Chr.8:143590454 - 143684444 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv466004 | Chr.8:143581491 - 143640022 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)