Assay Details
Target Gene Details
Entrez Gene ID: | 51005 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
CGI-14 |
Location: |
Chr.16:2520351-2531414 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMDHD2 | NM_001145815.1 | 11 | 1847 | NP_001139287.1 |
XM_017023262.1 | 11 | 2499 | XP_016878751.1 | |
XM_017023263.1 | 9 | 2487 | XP_016878752.1 | |
XM_017023264.1 | 10 | 2430 | XP_016878753.1 | |
XM_017023265.1 | 10 | 2397 | XP_016878754.1 | |
XM_017023266.1 | 10 | 2445 | XP_016878755.1 | |
XM_017023267.1 | 9 | 2441 | XP_016878756.1 | |
AK296877.1 | 11 | 1819 |
Target Gene Details
Entrez Gene ID: | 752014 |
Gene Name: | cementum protein 1 |
Gene Aliases: |
CP-23, CP23 |
Location: |
Chr.16:2530035-2531408 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEMP1 | NM_001048212.3 | 1 | 685 | NP_001041677.1 |
AY584596.1 | 1 | 685 | AAT01221.1 | |
BC146925.1 | 1 | 635 | ||
BC146926.1 | 1 | 635 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | SNORA78 CASKIN1 TBL3 IGFALS RAB11FIP3 NME4 ARHGDIG LMF1-AS1 UNKL TMEM8A TRAF7 LOC100134368 SYNGR3 CACNA1H STUB1 CAPN15 E4F1 SNHG9 NDUFB10 C16orf91 RPL3L PRSS27 MCRIP2 MIR3677 MPG LOC105371047 SNHG19 LOC105371046 RPL23AP5 IFT140 TSR3 EME2 PRSS29P MSRB1 RAB26 METRN TBC1D24 ATP6V0C LOC729652 PDIA2 RPS2 GNG13 HBZ GNPTG NOXO1 NHLRC4 NUBP2 KCTD5 RAB40C TPSD1 CCDC78 HN1L C1QTNF8 UBE2I DDX11L10 PKD1 PTX4 HBA2 CRAMP1 MIR3180-5 LUC7L CHTF18 TMEM204 HBQ1 PGP AMDHD2 MIR4516 ABCA17P MIR662 JMJD8 MRPL28 MIR6859-4 BAIAP3 MIR6767 LMF1 NME3 PIGQ PRR35 MIR6511B1 SNRNP25 WDR90 LINC00254 NTHL1 LOC652276 DNASE1L2 FAHD1 MIR5587 C16orf59 HBM ZNF598 LOC105371049 LOC105371045 RNF151 LOC106660606 TSC2 POLR3K LOC107984876 DECR2 RNPS1 MIR3176 METTL26 LOC105371038 FLJ42627 LOC100287175 CCNF MSLN MIR1225 RHOT2 HAGH FAM173A CEMP1 AXIN1 LINC00235 MAPK8IP3 BRICD5 ECI1 FAM234A MIR4717 CCDC154 SNORD60 MIR6768 LOC107987233 CLCN7 MRPS34 HS3ST6 SPSB3 MIR3177 WDR24 TPSAB1 WFIKKN1 NPRL3 NPW LOC105371184 NARFL MLST8 TELO2 NTN3 TPSG1 HBA1 RPUSD1 RGS11 GFER TPSB2 PRR25 FBXL16 MEIOB ABCA3 MIR3178 RHBDF1 SOX8 MIR940 SSTR5-AS1 WASIR2 SNORA10 SNORA64 SSTR5 SLC9A3R2 LOC101929280 ERVK13-1 HAGHL RHBDL1 PDPK1 |
esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | CEMP1 MIR3178 TCEB2 PRSS33 SRRM2 TBC1D24 ATP6V0C SRRM2-AS1 FLJ42627 PRSS21 ERVK13-1 PRSS27 PRSS41 LOC652276 KCTD5 PDPK1 AMDHD2 |
nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss | CEMP1 FLJ42627 MIR3178 ERVK13-1 LOC652276 TBC1D24 ATP6V0C PDPK1 AMDHD2 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | C16orf59 CASKIN1 LOC105371049 LOC106660606 TSC2 TRAF7 PKD1 MLST8 RNPS1 NTN3 E4F1 CCNF MIR3180-5 MIR1225 MIR3677 PGP AMDHD2 MIR4516 SNHG19 CEMP1 ABCA17P ABCA3 MIR3178 MIR940 BRICD5 RAB26 ECI1 MIR6767 MIR4717 TBC1D24 ATP6V0C SNORD60 MIR6768 LOC729652 MIR6511B1 LOC652276 DNASE1L2 PDPK1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |