Assay Details
Target Gene Details
Entrez Gene ID: | 5891 |
Gene Name: | MOK protein kinase |
Gene Aliases: |
RAGE, RAGE-1, RAGE1, STK30 |
Location: |
Chr.14:102224450-102305200 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOK | NM_001272011.1 | 11 | 1786 | NP_001258940.1 |
NM_014226.2 | 12 | 1876 | NP_055041.1 | |
NR_073540.1 | ||||
NR_073541.1 | 6 | 1156 | ||
NR_073542.1 | 5 | 1109 | ||
NR_073543.1 | 5 | 1072 | ||
XM_006720225.2 | XP_006720288.1 | |||
XM_006720226.2 | 10 | 1851 | XP_006720289.1 | |
XM_011537052.1 | XP_011535354.1 | |||
XM_011537053.2 | 5 | 1993 | XP_011535355.1 | |
XM_017021549.1 | 12 | 1873 | XP_016877038.1 | |
XM_017021553.1 | 10 | 1711 | XP_016877042.1 | |
XM_017021554.1 | 10 | 1710 | XP_016877043.1 | |
XM_017021555.1 | XP_016877044.1 | |||
XM_017021556.1 | XP_016877045.1 | |||
XM_017021557.1 | 9 | 1751 | XP_016877046.1 | |
XM_017021559.1 | 11 | 1865 | XP_016877048.1 | |
AB022694.1 | 12 | 1870 | BAA81688.1 | |
AK094003.1 | 2 | 2248 | ||
AK094747.1 | 9 | 2658 | ||
AK131542.1 | 2 | 3731 | ||
BC026069.1 | 11 | 2287 | AAH26069.1 | |
BC043179.1 | 10 | 2632 | AAH43179.1 | |
BC053536.1 | 11 | 2831 | AAH53536.1 | |
HY184670.1 | ||||
U46191.1 | 5 | 1049 | AAB38077.1 | |
U46192.1 | 5 | 1099 | AAB38080.1 | |
U46193.1 | 6 | 1156 | AAB38083.1 | |
U46194.1 | AAB38087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv22037 | Chr.14:102223226 - 102248955 on Build GRCh38 | Gain | WDR20 MOK |
nsv1429 | Chr.14:102201072 - 102246584 on Build GRCh38 | Deletion | WDR20 MOK |
esv3635547 | Chr.14:102223491 - 102317911 on Build GRCh38 | Gain | WDR20 MOK ZNF839 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |