Assay Details
Target Gene Details
Entrez Gene ID: | 84179 |
Gene Name: | major facilitator superfamily domain containing 7 |
Gene Aliases: |
LP2561 |
Location: |
Chr.4:676205-689468 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFSD7 | XM_005272299.4 | XP_005272356.1 |
Target Gene Details
Entrez Gene ID: | 4636 |
Gene Name: | myosin light chain 5 |
Gene Aliases: |
- |
Location: |
Chr.4:674199-682033 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYL5 | NM_002477.1 | NP_002468.1 | ||
XM_006713886.2 | XP_006713949.1 | |||
XM_017008245.1 | XP_016863734.1 | |||
XM_017008246.1 | XP_016863735.1 | |||
XM_017008247.1 | XP_016863736.1 | |||
XM_017008248.1 | XP_016863737.1 | |||
AK123974.1 | ||||
AK125529.1 | 4 | 1255 | ||
BC040050.1 | AAH40050.1 | |||
L03785.1 | AAA59890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv593206 | Chr.4:632173 - 741362 on Build GRCh38 | Loss | MFSD7 LOC101928521 MYL5 ATP5I PCGF3 PDE6B |
esv2759217 | Chr.4:609419 - 937566 on Build GRCh38 | Loss | MFSD7 LOC101928521 MYL5 ATP5I LOC100129917 CPLX1 LOC107986211 PCGF3 PDE6B GAK TMEM175 |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | CTBP1 MFSD7 LOC100129917 UVSSA CTBP1-AS CPLX1 LOC107986211 DGKQ NKX1-1 LOC100130872 LOC101928521 LOC105374344 ATP5I SLC26A1 MAEA CRIPAK TMED11P SPON2 CTBP1-AS2 PCGF3 FGFRL1 TMEM175 LOC101928548 MYL5 IDUA RNF212 PDE6B GAK |
nsv829834 | Chr.4:671688 - 781736 on Build GRCh38 | Loss | MFSD7 MYL5 ATP5I LOC100129917 LOC107986211 PCGF3 |
nsv950325 | Chr.4:620112 - 796912 on Build GRCh38 | Deletion | MFSD7 LOC101928521 MYL5 ATP5I LOC100129917 CPLX1 LOC107986211 PCGF3 PDE6B |
nsv593208 | Chr.4:674211 - 688444 on Build GRCh38 | Loss | MFSD7 MYL5 ATP5I |
nsv508986 | Chr.4:527899 - 756994 on Build GRCh38 | Insertion | MFSD7 LOC101928521 MYL5 ATP5I LOC107986211 PCGF3 PDE6B PIGG |
nsv10413 | Chr.4:673415 - 690512 on Build GRCh38 | Gain | MFSD7 MYL5 ATP5I |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |