Assay Details
Target Gene Details
Entrez Gene ID: | 55809 |
Gene Name: | transcriptional regulating factor 1 |
Gene Aliases: |
BCAR2, HSA277276, RAPA, TREP132, TReP-132, dJ139D8.5 |
Location: |
Chr.6:42224933-42452216 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830649 | Chr.6:42047596 - 42231697 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)