Assay Details
Target Gene Details
Entrez Gene ID: | 8312 |
Gene Name: | axin 1 |
Gene Aliases: |
AXIN, PPP1R49 |
Location: |
Chr.16:287440-355226 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AXIN1 | NM_003502.3 | NP_003493.1 | ||
NM_181050.2 | NP_851393.1 | |||
NR_134879.1 | ||||
XM_011522682.2 | XP_011520984.1 | |||
XM_011522683.2 | XP_011520985.1 | |||
XM_011522684.2 | XP_011520986.1 | |||
XM_011522686.1 | XP_011520988.1 | |||
XM_017023743.1 | XP_016879232.1 | |||
XM_017023744.1 | XP_016879233.1 | |||
XM_017023745.1 | XP_016879234.1 | |||
XM_017023746.1 | XP_016879235.1 | |||
AB208876.1 | BAD92113.1 | |||
AF009674.1 | AAC51624.1 | |||
BC017447.1 | AAH17447.1 | |||
BC044648.1 | AAH44648.1 | |||
BX648449.1 | ||||
KF615847.1 | ||||
KF615848.1 | ||||
KF615849.1 | ||||
KF615850.1 | ||||
KF615851.1 | ||||
KF615852.1 | ||||
KF615853.1 | ||||
KF615854.1 | ||||
KU178413.1 | ||||
KU178414.1 | ||||
KU178415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | PIGQ LOC107984876 WDR24 LMF1 RPL3L LOC101929280 MIR4717 MAPK8IP3 CCDC154 CRAMP1 SLC9A3R2 PTX4 LOC107987233 EME2 BRICD5 TPSG1 LUC7L PDPK1 NPW LOC106660606 TRAF7 WASIR2 C1QTNF8 SNRNP25 LOC105371049 LOC100134368 TSR3 TBC1D24 DDX11L10 TPSAB1 PRSS27 PDIA2 CASKIN1 MIR3176 AXIN1 HAGH WFIKKN1 FAHD1 CHTF18 SNORA64 TPSB2 NHLRC4 RGS11 TBL3 ATP6V0C MEIOB NME3 GFER FAM234A LINC00235 CEMP1 HN1L LOC105371038 PRR35 NOXO1 ERVK13-1 MCRIP2 DNASE1L2 LOC105371047 WDR90 C16orf59 E4F1 RAB11FIP3 CLCN7 MIR1225 METTL26 SNHG19 HBZ MIR6767 MRPS34 NTHL1 TELO2 SPSB3 NPRL3 MIR3178 FAM173A TMEM8A NUBP2 ARHGDIG SSTR5 HBA1 CACNA1H RPS2 MIR662 TSC2 CAPN15 TPSD1 LOC105371045 GNG13 LOC652276 SYNGR3 FLJ42627 AMDHD2 IGFALS UNKL CCDC78 NME4 MIR3677 MPG MIR5587 PRSS29P MSRB1 PKD1 RPL23AP5 C16orf91 LOC105371046 MIR3180-5 PGP LMF1-AS1 TMEM204 HS3ST6 RNF151 MLST8 NDUFB10 MIR6511B1 MIR940 LINC00254 RHOT2 MRPL28 JMJD8 NTN3 PRR25 SSTR5-AS1 HBQ1 ECI1 FBXL16 RPUSD1 MIR3177 SNHG9 RAB26 UBE2I HAGHL ABCA3 LOC105371184 MIR4516 ABCA17P BAIAP3 MIR6768 STUB1 KCTD5 LOC729652 LOC100287175 SNORA10 POLR3K HBM IFT140 RHBDL1 RAB40C DECR2 SNORA78 NARFL SNORD60 RHBDF1 MIR6859-4 ZNF598 HBA2 CCNF SOX8 RNPS1 METRN GNPTG MSLN |
esv3637564 | Chr.16:287941 - 370218 on Build GRCh38 | Gain | MRPL28 AXIN1 |
nsv952896 | Chr.16:273501 - 321500 on Build GRCh38 | Deletion | ARHGDIG RGS11 PDIA2 AXIN1 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RPL23AP5 TMEM8A ARHGDIG LOC100134368 RGS11 NME4 PDIA2 FAM234A RAB11FIP3 MRPL28 AXIN1 DECR2 |
nsv570951 | Chr.16:279671 - 322597 on Build GRCh38 | Loss | ARHGDIG PDIA2 AXIN1 |
nsv827477 | Chr.16:262247 - 297639 on Build GRCh38 | Gain | ARHGDIG RGS11 PDIA2 FAM234A AXIN1 |
nsv570947 | Chr.16:272935 - 297326 on Build GRCh38 | Loss | ARHGDIG RGS11 PDIA2 AXIN1 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RPL23AP5 TMEM8A ARHGDIG LOC100134368 RGS11 NME4 PDIA2 FAM234A RAB11FIP3 MRPL28 AXIN1 DECR2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |