Assay Details
Target Gene Details
Entrez Gene ID: | 93377 |
Gene Name: | oligodendrocytic myelin paranodal and inner loop protein |
Gene Aliases: |
HTMP10, TMEM10, TMP10 |
Location: |
Chr.10:96343216-96359365 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OPALIN | NM_001040102.2 | 4 | 3177 | NP_001035191.1 |
NM_001040103.2 | 7 | 3285 | NP_001035192.1 | |
NM_001284320.1 | 5 | 3213 | NP_001271249.1 | |
NM_001284321.1 | 5 | 3210 | NP_001271250.1 | |
NM_001284322.1 | 7 | 3328 | NP_001271251.1 | |
NM_001284323.1 | 7 | 3063 | NP_001271252.1 | |
NM_001284324.1 | 7 | 2956 | NP_001271253.1 | |
NM_001284326.1 | 6 | 3024 | NP_001271255.1 | |
NM_001284327.1 | 6 | 2917 | NP_001271256.1 | |
NM_033207.4 | 6 | 3246 | NP_149984.1 | |
NR_104296.1 | 6 | 3305 | ||
NR_104297.1 | 6 | 3232 | ||
AF367761.2 | 6 | 3186 | AAK62349.1 | |
AL834229.1 | 6 | 2863 | CAD38907.1 | |
BC033737.1 | 6 | 2863 | AAH33737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831950 | Chr.10:96321215 - 96485677 on Build GRCh38 | Gain | DNTT TLL2 OPALIN |
nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain | DNTT TLL2 BLNK OPALIN |