Assay Details
Target Gene Details
Entrez Gene ID: | 80380 |
Gene Name: | programmed cell death 1 ligand 2 |
Gene Aliases: |
B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2 |
Location: |
Chr.9:5510438-5571282 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDCD1LG2 | NM_025239.3 | 4 | 761 | NP_079515.2 |
XM_005251600.3 | 4 | 868 | XP_005251657.1 | |
AF329193.1 | 4 | 746 | AAK31105.1 | |
AF344424.1 | 4 | 761 | AAK15370.1 | |
AK001872.1 | 2 | 192 | ||
AY254343.1 | 3 | 488 | AAP13471.1 | |
AY271901.1 | AAP49000.1 | |||
AY271902.1 | AAP49001.1 | |||
BC074766.2 | 4 | 522 | AAH74766.1 | |
BC113678.1 | 4 | 524 | ||
BC113680.1 | 4 | 524 | ||
BX951459.1 | 2 | 398 | ||
DQ336699.1 | 3 | 455 | ABC59616.1 | |
EF444806.1 | 4 | 608 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |