Assay Details
Target Gene Details
Entrez Gene ID: | 283232 |
Gene Name: | transmembrane protein 80 |
Gene Aliases: |
- |
Location: |
Chr.11:695386-705028 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM80 | NM_001042463.2 | NP_001035928.2 | ||
NM_001276253.1 | NP_001263182.1 | |||
NM_001276274.1 | NP_001263203.1 | |||
NM_174940.3 | NP_777600.3 | |||
XM_006718206.3 | XP_006718269.3 | |||
XM_017017600.1 | XP_016873089.1 | |||
AK124309.1 | ||||
AK302585.1 | ||||
BC008671.1 | ||||
BI916171.1 | ||||
BM874316.1 | ||||
CF134831.1 | ||||
CK819789.1 | ||||
DR006597.1 | ||||
HY080028.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625074 | Chr.11:669311 - 702135 on Build GRCh38 | Loss | TMEM80 DEAF1 |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss | LOC101927503 MIR210HG SLC25A22 MUC6 SNORA52 PHRF1 POLR2L DEAF1 SCT MUC2 MIR6744 PNPLA2 PTDSS2 MUC5AC LOC171391 IRF7 TOLLIP AP2A2 RNH1 PDDC1 TSPAN4 CRACR2B ANO9 LOC143666 SIGIRR LMNTD2 CHID1 PIDD1 B4GALNT4 PKP3 PANO1 CEND1 TMEM80 RPLP2 MUC5B CD151 EPS8L2 LOC101059906 DRD4 CDHR5 RASSF7 TALDO1 HRAS LRRC56 MIR210 |
nsv832043 | Chr.11:631027 - 758602 on Build GRCh38 | Loss | TMEM80 DEAF1 TALDO1 EPS8L2 DRD4 |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss | LOC101927503 MIR210HG SLC25A22 MUC6 SNORA52 PHRF1 POLR2L DEAF1 SCT MUC2 MIR6744 PNPLA2 PTDSS2 MUC5AC LOC171391 TOLLIP-AS1 IRF7 TOLLIP AP2A2 RNH1 PDDC1 TSPAN4 CRACR2B ANO9 LOC143666 SIGIRR LMNTD2 CHID1 PIDD1 PANO1 CEND1 TMEM80 RPLP2 MUC5B CD151 EPS8L2 LOC101059906 DRD4 CDHR5 RASSF7 TALDO1 HRAS LRRC56 MIR210 |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion | MIR210HG SLC25A22 SNORA52 PHRF1 POLR2L DEAF1 SCT PNPLA2 PTDSS2 LOC171391 IRF7 AP2A2 RNH1 PDDC1 TSPAN4 CRACR2B ANO9 LOC143666 SIGIRR LMNTD2 CHID1 PIDD1 B4GALNT4 PKP3 PANO1 CEND1 TMEM80 RPLP2 CD151 EPS8L2 LOC101059906 DRD4 CDHR5 RASSF7 TALDO1 HRAS LRRC56 MIR210 |
nsv1159791 | Chr.11:461793 - 909492 on Build GRCh38 | Deletion | MIR210HG SLC25A22 SNORA52 PHRF1 POLR2L DEAF1 SCT PNPLA2 PTDSS2 LOC171391 IRF7 RNH1 PDDC1 TSPAN4 CRACR2B LOC143666 LMNTD2 CHID1 PIDD1 PANO1 CEND1 TMEM80 RPLP2 CD151 EPS8L2 LOC101059906 DRD4 CDHR5 RASSF7 TALDO1 HRAS LRRC56 MIR210 |