Assay Details
Target Gene Details
Entrez Gene ID: | 105370792 |
Gene Name: | uncharacterized LOC105370792 |
Gene Aliases: |
- |
Location: |
Chr.15:41892777-41898815 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105370792 | NR_135681.1 | |||
AK057625.1 | 1 | 1188 | ||
DA112916.1 |
Target Gene Details
Entrez Gene ID: | 51332 |
Gene Name: | spectrin beta, non-erythrocytic 5 |
Gene Aliases: |
BSPECV, HUBSPECV, HUSPECV |
Location: |
Chr.15:41848146-41894077 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPTBN5 | NM_016642.3 | 1 | 98 | NP_057726.4 |
AF233523.1 | 1 | 98 | AAF65317.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9244 | Chr.15:41818514 - 41916607 on Build GRCh38 | Gain | EHD4 JMJD7 MAPKBP1 JMJD7-PLA2G4B PLA2G4B LOC105370792 MIR4310 SPTBN5 |
nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain | MAPKBP1 PLA2G4D PLA2G4E EHD4-AS1 PLA2G4B EHD4 JMJD7 JMJD7-PLA2G4B PLA2G4E-AS1 LOC105370792 PLA2G4F MIR4310 SPTBN5 |
nsv1038933 | Chr.15:41846258 - 42016889 on Build GRCh38 | Gain | EHD4 JMJD7-PLA2G4B PLA2G4E EHD4-AS1 PLA2G4E-AS1 PLA2G4B LOC105370792 MIR4310 SPTBN5 |
nsv1050892 | Chr.15:41836657 - 41894284 on Build GRCh38 | Gain | JMJD7 JMJD7-PLA2G4B PLA2G4B LOC105370792 MIR4310 SPTBN5 |
nsv517631 | Chr.15:41804728 - 41895294 on Build GRCh38 | Loss | JMJD7 MAPKBP1 JMJD7-PLA2G4B PLA2G4B LOC105370792 MIR4310 SPTBN5 |
dgv2582n100 | Chr.15:41791376 - 41900562 on Build GRCh38 | Gain | EHD4 JMJD7 MAPKBP1 JMJD7-PLA2G4B PLA2G4B LOC105370792 MIR4310 SPTBN5 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |