Assay Details
Target Gene Details
Entrez Gene ID: | 6886 |
Gene Name: | TAL bHLH transcription factor 1, erythroid differentiation factor |
Gene Aliases: |
SCL, TCL5, bHLHa17, tal-1 |
Location: |
Chr.1:47216290-47232389 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TAL1 | NM_001287347.2 | 5 | 4180 | NP_001274276.1 |
NM_001290403.1 | 5 | 4290 | NP_001277332.1 | |
NM_001290404.1 | 6 | 4295 | NP_001277333.1 | |
NM_001290405.1 | 5 | 4163 | NP_001277334.1 | |
NM_001290406.1 | 3 | 3687 | NP_001277335.1 | |
NM_003189.5 | 6 | 4408 | NP_003180.1 | |
XM_005271160.4 | 5 | 5790 | XP_005271217.1 | |
XM_017002187.1 | 5 | 5205 | XP_016857676.1 | |
XM_017002188.1 | 5 | 4397 | XP_016857677.1 | |
XM_017002189.1 | 5 | 4200 | XP_016857678.1 | |
XM_017002190.1 | 5 | 4896 | XP_016857679.1 | |
XM_017002191.1 | 4 | 4021 | XP_016857680.1 | |
XM_017002193.1 | 4 | 3757 | XP_016857682.1 | |
BQ186967.1 | 1 | 59 | ||
M61108.1 | 4 | 4541 | AAA36600.1 | |
S53245.1 | 5 | 4180 | AAB19683.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75623452] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |