Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | SLC38A10 LOC100129503 FAAP100 TSPAN10 AATK LOC107985021 BAHCC1 FSCN2 NPLOC4 LOC107985022 MIR6786 CHMP6 LOC400627 LINC00482 TMEM105 LOC100130370 MIR657 AATK-AS1 LOC105371925 LOC101928855 OXLD1 BAIAP2-AS1 BAIAP2 MIR3065 MIR4740 ARL16 MIR3186 CCDC137 PDE6G TEPSIN SLC25A10 HGS LOC105371921 MIR1250 C17orf89 RPTOR CEP131 MRPL12 MIR338 ACTG1 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | AATK-AS1 AATK CEP131 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | SLC38A10 FAAP100 HEXDC FSCN2 NPLOC4 MIR6786 FN3K FN3KRP AATK-AS1 CSNK1D OGFOD3 FASN ARHGDIA TEX19 MAFG MIR4740 LOC105376790 MIR3186 STRA13 WDR45B SECTM1 CCDC137 B3GNTL1 PDE6G TEPSIN SNORD134 MIR4525 DCXR C17orf89 PCYT2 CEP131 P4HB PPP1R27 MRPL12 GCGR NPB FOXK2 ALYREF MIR6787 TSPAN10 MCRIP1 SIRT7 AATK BAHCC1 RFNG RAC3 LRRC45 LINC00482 DUS1L TMEM105 LOC100130370 MYADML2 LOC105371944 LOC105371925 CD7 OXLD1 ASPSCR1 TBCD MAFG-AS1 ARL16 C17orf62 PYCR1 RAB40B UTS2R GPS1 ZNF750 SLC25A10 LOC101929552 NOTUM CCDC57 HGS NARF ANAPC11 LOC101929511 LOC105376791 SLC16A3 ACTG1 |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | SLC38A10 C17orf89 AATK-AS1 LOC105371925 AATK CEP131 TEPSIN |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | MIR1250 MIR657 AATK-AS1 AATK CEP131 BAIAP2 MIR3065 MIR338 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | SLC38A10 AATK TEPSIN LINC00482 TMEM105 MIR1250 C17orf89 MIR657 AATK-AS1 LOC105371925 BAIAP2-AS1 CEP131 BAIAP2 MIR3065 MIR338 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR1250 MIR657 AATK-AS1 AATK BAIAP2-AS1 CEP131 BAIAP2 MIR3065 MIR338 |
nsv1064123 | Chr.17:81168449 - 81282457 on Build GRCh38 | Gain | SLC38A10 C17orf89 AATK-AS1 LOC105371925 CEP131 TEPSIN |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | SLC38A10 MIR3186 FAAP100 AATK BAHCC1 FSCN2 NPLOC4 TEPSIN LINC00482 TMEM105 LOC100130370 C17orf89 AATK-AS1 LOC105371925 CEP131 MIR4740 ACTG1 |