Assay Details
Target Gene Details
Entrez Gene ID: | 84168 |
Gene Name: | anthrax toxin receptor 1 |
Gene Aliases: |
ATR, GAPO, TEM8 |
Location: |
Chr.2:69013144-69249327 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANTXR1 | NM_018153.3 | 1 | 399 | NP_060623.2 |
NM_032208.2 | 1 | 399 | NP_115584.1 | |
NM_053034.2 | 1 | 399 | NP_444262.1 | |
XM_017005075.1 | 1 | 190 | XP_016860564.1 | |
XM_017005076.1 | 1 | 190 | XP_016860565.1 | |
XM_017005077.1 | 1 | 190 | XP_016860566.1 | |
AF279145.2 | 1 | 186 | AAK52094.1 | |
AF421380.1 | 1 | 146 | AAL26496.1 | |
AK001463.1 | 1 | 186 | BAA91707.1 | |
AK223273.1 | 1 | 173 | BAD96993.1 | |
AK292113.1 | 1 | 121 | ||
AK309553.1 | 1 | 152 | ||
BC012074.1 | 1 | 155 | AAH12074.1 | |
CF126791.1 | 1 | 280 | ||
DC360415.1 | 1 | 396 | ||
DC361072.1 | 1 | 422 | ||
JX424838.1 | 1 | 166 | ||
JX424839.1 | 1 | 166 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |