Assay Details
Target Gene Details
Entrez Gene ID: | 27158 |
Gene Name: | NADPH dependent diflavin oxidoreductase 1 |
Gene Aliases: |
NR1, bA350O14.9 |
Location: |
Chr.9:137205667-137219361 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDOR1 | NM_001144026.2 | 8 | 1114 | NP_001137498.1 |
NM_001144027.2 | 7 | 1012 | NP_001137499.1 | |
NM_001144028.2 | 8 | 1114 | NP_001137500.1 | |
NM_014434.3 | 8 | 1114 | NP_055249.1 | |
XM_011518545.1 | 7 | 1002 | XP_011516847.1 | |
XM_011518547.1 | 8 | 904 | XP_011516849.1 | |
XM_011518548.2 | 7 | 2304 | XP_011516850.1 | |
XM_017014626.1 | 8 | 1114 | XP_016870115.1 | |
XM_017014627.1 | 8 | 1114 | XP_016870116.1 | |
AF199509.1 | 8 | 1050 | AAF25205.1 | |
AK026089.1 | 8 | 1086 | ||
AK074403.1 | 7 | 973 | ||
AK225995.1 | 8 | 1085 | ||
AK290026.1 | 8 | 1111 | ||
AY077845.1 | 8 | 1032 | AAL77754.1 | |
BC015735.1 | 8 | 1050 | AAH15735.1 | |
BC093782.1 | 8 | 1031 | AAH93782.1 | |
BC111943.1 | 8 | 1031 | AAI11944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv13010n54 | Chr.9:137191539 - 137233413 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv616012 | Chr.9:137197680 - 137223834 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv616007 | Chr.9:137079368 - 137240666 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)