Assay Details
Target Gene Details
Entrez Gene ID: | 56474 |
Gene Name: | CTP synthase 2 |
Gene Aliases: |
- |
Location: |
Chr.X:16587999-16712979 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTPS2 | NM_001144002.1 | NP_001137474.1 | ||
NM_019857.4 | NP_062831.3 | |||
NM_175859.2 | NP_787055.1 | |||
XM_005274562.3 | XP_005274619.1 | |||
XM_006724503.3 | XP_006724566.1 | |||
XM_011545545.2 | XP_011543847.1 | |||
AF086422.1 | ||||
AF226667.1 | AAF91241.1 | |||
AK023549.1 | BAB14607.1 | |||
AK024070.1 | BAB14814.1 | |||
AK025654.1 | BAB15204.1 | |||
AK125332.1 | ||||
AK125348.1 | ||||
AK309508.1 | ||||
BC006256.2 | AAH06256.2 | |||
BC034986.1 | AAH34986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6815 | Chr.X:16609172 - 16640869 on Build GRCh38 | Insertion | CTPS2 MIR548AM |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |