Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | LOC107986711 SNORA14A TMEM120A POR STYXL1 GTF2IP7 MDH2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | ZP3 GTF2IRD2B SRRM3 STAG3L1 SPDYE16 MIR4651 YWHAG LOC105375355 GTF2IP7 NCF1C CCL24 TMEM120A SPDYE5 PMS2P3 HSPB1 LOC441259 TRIM73 RHBDD2 LOC107986711 GTF2IP1 POM121C FDPSP2 PMS2P10 LOC100133091 POR CCL26 DTX2 SSC4D NSUN5P1 POMZP3 SNORA14A HIP1 STYXL1 PMS2P2 LOC541473 MDH2 |
esv3613746 | Chr.7:75965095 - 76003876 on Build GRCh38 | Loss | TMEM120A POR STYXL1 |
esv3613748 | Chr.7:75992925 - 75999800 on Build GRCh38 | Loss | TMEM120A STYXL1 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | SNORA14A TMEM120A MIR4651 POR STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | ZP3 SRRM3 SPDYE16 MIR4651 YWHAG LOC105375355 GTF2IP7 CCL24 TMEM120A SPDYE5 PMS2P3 HSPB1 RHBDD2 LOC107986711 POM121C FDPSP2 LOC100133091 POR CCL26 DTX2 SSC4D POMZP3 SNORA14A HIP1 STYXL1 MDH2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |