Assay Details
Target Gene Details
Entrez Gene ID: | 65220 |
Gene Name: | NAD kinase |
Gene Aliases: |
dJ283E3.1 |
Location: |
Chr.1:1751232-1780514 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NADK | NM_001198993.1 | 4 | 502 | NP_001185922.1 |
NM_001198994.1 | 6 | 866 | NP_001185923.1 | |
NM_001198995.1 | 2 | 364 | NP_001185924.1 | |
NM_023018.4 | 4 | 571 | NP_075394.3 | |
XM_005244778.2 | 4 | 632 | XP_005244835.1 | |
XM_006710837.2 | 5 | 747 | XP_006710900.1 | |
XM_006710838.2 | 4 | 480 | XP_006710901.1 | |
XM_006710839.1 | 7 | 1059 | XP_006710902.1 | |
XM_011542006.1 | 4 | 553 | XP_011540308.1 | |
XM_011542007.1 | 4 | 632 | XP_011540309.1 | |
XM_011542008.1 | 4 | 490 | XP_011540310.1 | |
XM_011542009.2 | 3 | 381 | XP_011540311.1 | |
XM_017002143.1 | 4 | 632 | XP_016857632.1 | |
XM_017002144.1 | 2 | 376 | XP_016857633.1 | |
XM_017002145.1 | 4 | 541 | XP_016857634.1 | |
XM_017002146.1 | 4 | 411 | XP_016857635.1 | |
AF250320.1 | 6 | 866 | AAG44568.1 | |
AK023114.1 | 4 | 571 | BAB14412.1 | |
AK290161.1 | 4 | 553 | ||
AK291599.1 | 4 | 495 | ||
AK291728.1 | 4 | 471 | ||
AK296722.1 | 2 | 364 | ||
AK307645.1 | 3 | 353 | ||
AY090771.1 | 3 | 349 | AAM01195.1 | |
BC001709.1 | 4 | 434 | AAH01709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003545 | Chr.1:1696467 - 1760882 on Build GRCh38 | Gain+Loss | SLC35E2 CDK11A MMP23A NADK |
dgv5n111 | Chr.1:1575421 - 1823403 on Build GRCh38 | Deletion | SLC35E2B SLC35E2 MMP23B CDK11A GNB1 MMP23A FNDC10 NADK MIB2 CDK11B |
nsv834091 | Chr.1:1701359 - 1880638 on Build GRCh38 | Loss | SLC35E2 CDK11A GNB1 MMP23A NADK |
nsv545041 | Chr.1:1753033 - 1780259 on Build GRCh38 | Loss | NADK |
nsv945749 | Chr.1:1751857 - 1760427 on Build GRCh38 | Duplication | NADK |
nsv545030 | Chr.1:1689422 - 1774697 on Build GRCh38 | Gain | SLC35E2B SLC35E2 CDK11A MMP23A NADK |
nsv10161 | Chr.1:776731 - 1777210 on Build GRCh38 | Gain+Loss | CPSF3L LINC01128 ISG15 LOC100130417 CPTP RNF223 TMEM240 SLC35E2 LOC107985728 HES4 AURKAIP1 TNFRSF4 MIR6808 LOC284600 C1orf159 KLHL17 PERM1 ATAD3A TTLL10-AS1 MMP23B ANKRD65 LOC105378948 ACAP3 SSU72 SAMD11 LOC102724312 B3GALT6 MMP23A LOC148413 LOC100288175 PUSL1 MRPL20 TNFRSF18 CCNL2 FAM87B MIB2 CDK11B MXRA8 UBE2J2 FAM132A LOC107985729 MIR200A LOC100287934 TTLL10 CDK11A ATAD3C LINC01342 FAM41C MIR429 TAS1R3 SLC35E2B AGRN FNDC10 SCNN1D NOC2L ATAD3B MIR200B MIR6727 LOC100288069 DVL1 TMEM88B PLEKHN1 NADK VWA1 SDF4 MIR6726 LINC00115 |
nsv1160797 | Chr.1:1566216 - 1805918 on Build GRCh38 | Duplication | SLC35E2B SLC35E2 MMP23B CDK11A GNB1 MMP23A FNDC10 NADK SSU72 MIB2 CDK11B |
nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss | LOC100132287 CPSF3L MORN1 WASH7P LOC101928626 RNF223 TMEM240 SLC35E2 LOC107985728 HES4 LOC729737 KLHL17 ATAD3A OR4F5 MMP23B FAAP20 ANKRD65 ACAP3 SSU72 SAMD11 LOC100506504 GABRD LOC102724312 MTND1P23 MMP23A LOC105378589 LOC148413 PUSL1 FAM87B MIB2 TMEM52 UBE2J2 LOC105378949 OR4F16 CDK11A ATAD3C SLC35E2B AGRN FNDC10 SCNN1D MIR200B MIR6859-2 MIR6727 DVL1 MIR6859-1 PLEKHN1 NADK SDF4 MIR6726 LINC00115 OR4F29 LOC107984841 LINC01128 SKI PRKCZ ISG15 LOC105378592 LOC100130417 CPTP AURKAIP1 TNFRSF4 MIR6808 LOC284600 C1orf159 LOC100129534 PERM1 TTLL10-AS1 LOC105378948 MIR1302-2 LOC105378591 CALML6 B3GALT6 LOC100288175 MRPL20 TNFRSF18 CCNL2 CDK11B MXRA8 LOC105378947 LOC102725121 FAM132A FAM138A LOC107985729 MIR200A LOC100287934 TTLL10 LINC01342 FAM41C MIR429 TAS1R3 MIR6723 GNB1 LOC100134822 NOC2L ATAD3B LOC100288069 LOC100133331 TMEM88B CFAP74 MTND2P28 DDX11L1 VWA1 |
nsv950521 | Chr.1:1729062 - 1760461 on Build GRCh38 | Deletion | SLC35E2 NADK |
esv2758913 | Chr.1:1389149 - 2092899 on Build GRCh38 | Gain+Loss | TMEM52 LOC105378949 LOC107985729 PRKCZ TMEM240 SLC35E2 CDK11A ATAD3C SLC35E2B ATAD3A MMP23B GNB1 ANKRD65 FNDC10 SSU72 GABRD LOC105378591 ATAD3B CALML6 LOC102724312 TMEM88B CFAP74 MMP23A LOC105378589 LOC148413 NADK VWA1 MRPL20 CCNL2 MIB2 CDK11B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |