Assay Details
Target Gene Details
Entrez Gene ID: | 79143 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7 |
Location: |
Chr.19:54173412-54190050 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT7 | NM_001146056.2 | 6 | 2199 | NP_001139528.1 |
NM_001146083.2 | 7 | 2278 | NP_001139555.1 | |
NM_024298.4 | 8 | 2405 | NP_077274.3 | |
XM_011527299.2 | 8 | 2609 | XP_011525601.1 | |
XM_011527300.2 | 8 | 2000 | XP_011525602.1 | |
AF211969.1 | 2 | 860 | ||
AI032483.1 | 1 | 194 | ||
AK055908.1 | 8 | 2338 | BAB71043.1 | |
AK122789.1 | 9 | 2123 | ||
AK123290.1 | 2 | 2852 | ||
AK298689.1 | 6 | 1885 | ||
AW014416.1 | 1 | 211 | ||
BC002512.2 | 7 | 1855 | AAH02512.2 | |
BC003164.1 | 8 | 1936 | AAH03164.2 | |
BC015857.1 | 7 | 1842 | ||
BQ045276.1 | 1 | 212 | ||
S82470.1 | 6 | 1711 | AAB37433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain | TMC4 LENG1 MBOAT7 TSEN34 |
nsv515623 | Chr.19:54102901 - 54179150 on Build GRCh38 | Loss | CNOT3 PRPF31 TMC4 LENG1 NDUFA3 MBOAT7 TFPT |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | CNOT3 PRPF31 TMC4 LENG1 NDUFA3 MBOAT7 TFPT TSEN34 OSCAR |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | MIR516B1 MIR518E MIR520G MIR525 TMC4 ZNF331 MIR372 MIR518C TPM3P9 MBOAT7 NLRP12 TFPT MIR526A2 VN1R4 MIR516A2 MIR516B2 LOC284379 MIR517A LILRA5 ZNF525 MIR520C VN1R2 PRPF31 MIR518B NDUFA3 MIR1323 MIR523 ZNF818P MIR519B CNOT3 MIR519C MIR520D TARM1 MIR517B VSTM1 MIR519A1 MYADM TSEN34 OSCAR MIR515-1 ZNF765 MIR1283-1 MIR524 ZNF677 MIR526B LENG1 MIR520H PRKCG CACNG7 MIR517C MIR515-2 MIR516A1 MIR519E MIR521-2 ZNF845 ZNF761 RPS9 MIR519D MIR520A DPRX FAM90A27P MIR373 MIR521-1 ZNF665 CACNG6 MIR518A1 BIRC8 MIR518A2 MIR527 LILRB2 MIR518F LILRA6 CACNG8 MIR519A2 MIR522 MIR520B MIR518D LOC105372457 LILRB5 MIR526A1 MIR520E LILRB3 MIR512-1 MIR512-2 MIR935 MIR371B MIR520F MIR1283-2 MIR371A MIR4752 ZNF813 MIR498 |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion | LILRA6 TMC4 LENG1 LILRB5 MBOAT7 LILRB3 TSEN34 RPS9 |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss | LILRA2 TMC4 KIR3DL2 KIR2DL1 LILRB4 KIR3DL3 MBOAT7 RPS9 LENG9 LILRA5 LILRA1 KIR2DL4 TTYH1 LENG8-AS1 MIR8061 CDC42EP5 KIR3DL1 LILRP2 KIR3DX1 FCAR LOC101928804 LILRB1 LILRA4 LILRB2 LOC102724300 LILRA6 LILRB5 LILRB3 LAIR1 TSEN34 LAIR2 NCR1 LENG8 KIR2DS4 KIR2DL3 MIR4752 NLRP7 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |