Assay Details
Target Gene Details
Entrez Gene ID: | 79623 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 14 |
Gene Aliases: |
GALNT15, GalNac-T10, GalNac-T14 |
Location: |
Chr.2:30888518-31138726 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT14 | NM_001253826.1 | 16 | 2421 | NP_001240755.1 |
NM_001253827.1 | 17 | 2186 | NP_001240756.1 | |
NM_024572.3 | 15 | 2406 | NP_078848.2 | |
XM_011533105.1 | 17 | 1940 | XP_011531407.1 | |
XM_017004906.1 | 15 | 1871 | XP_016860395.1 | |
XM_017004907.1 | 13 | 1653 | XP_016860396.1 | |
XM_017004908.1 | 13 | 1936 | XP_016860397.1 | |
AK022753.1 | 16 | 1842 | BAB14227.1 | |
AK074394.1 | 9 | 1066 | ||
AK091313.1 | 15 | 1776 | BAC03634.1 | |
AK122747.1 | 17 | 2170 | ||
AK225808.1 | 15 | 2129 | ||
AK298735.1 | ||||
AY358758.1 | 15 | 2099 | AAQ89118.1 | |
BC006269.2 | 8 | 975 | AAH06269.2 | |
BC010659.1 | 15 | 2406 | AAH10659.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833659 | Chr.2:30797706 - 30948639 on Build GRCh38 | Gain | GALNT14 CAPN13 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |