Assay Details
Target Gene Details
Entrez Gene ID: | 11336 |
Gene Name: | exocyst complex component 3 |
Gene Aliases: |
SEC6, SEC6L1, Sec6p |
Location: |
Chr.5:443184-471938 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3 | NM_007277.4 | 4 | 1105 | NP_009208.2 |
XM_011513948.2 | 4 | 1140 | XP_011512250.1 | |
XM_011513949.2 | 4 | 1157 | XP_011512251.1 | |
XM_011513950.2 | 3 | 1035 | XP_011512252.1 | |
XM_011513951.2 | 4 | 1134 | XP_011512253.1 | |
XM_017009000.1 | 4 | 1140 | XP_016864489.1 | |
XM_017009001.1 | 4 | 1140 | XP_016864490.1 | |
XM_017009002.1 | 4 | 1140 | XP_016864491.1 | |
XM_017009003.1 | 4 | 1140 | XP_016864492.1 | |
XM_017009004.1 | 5 | 1218 | XP_016864493.1 | |
XM_017009005.1 | 4 | 1134 | XP_016864494.1 | |
XM_017009006.1 | 4 | 1134 | XP_016864495.1 | |
AF055006.1 | 1 | 157 | AAC09358.1 | |
AK074086.1 | 3 | 1096 | BAB84912.1 | |
AK131103.1 | 1 | 586 | BAC85153.1 | |
AK315749.1 | 1 | 613 | ||
AL832434.1 | 2 | 761 | CAH10613.1 | |
BC001511.2 | 1 | 517 | AAH01511.2 | |
BC019304.1 | 2 | 687 | AAH19304.1 | |
BC064569.1 | 3 | 1047 | AAH64569.1 | |
GU727644.1 | 4 | 1105 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894107 | Chr.5:81322 - 801057 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv830183 | Chr.5:349237 - 502409 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3603748 | Chr.5:453680 - 475646 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1032584 | Chr.5:375290 - 809679 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv596626 | Chr.5:429874 - 598109 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1030767 | Chr.5:154674 - 457840 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv461890 | Chr.5:438449 - 539195 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2759315 | Chr.5:17263 - 595840 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3603744 | Chr.5:359572 - 478120 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2752071 | Chr.5:378298 - 699075 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1019215 | Chr.5:346521 - 661696 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1027279 | Chr.5:337575 - 531110 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3603745 | Chr.5:376684 - 458243 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)