Assay Details
Target Gene Details
Entrez Gene ID: | 2890 |
Gene Name: | glutamate ionotropic receptor AMPA type subunit 1 |
Gene Aliases: |
GLUH1, GLUR1, GLURA, GluA1, HBGR1 |
Location: |
Chr.5:153489615-153813873 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIA1 | NM_000827.3 | 1 | 377 | NP_000818.2 |
NM_001114183.1 | 1 | 377 | NP_001107655.1 | |
NM_001258019.1 | 1 | 377 | NP_001244948.1 | |
NM_001258020.1 | 1 | 377 | NP_001244949.1 | |
NR_047578.1 | 1 | 377 | ||
XM_011537635.2 | XP_011535937.1 | |||
AB209094.1 | 1 | 240 | BAD92331.1 | |
AK289565.1 | ||||
AK294011.1 | 1 | 231 | ||
AK295827.1 | 1 | 246 | ||
AK310189.1 | ||||
BC111734.1 | 1 | 102 | AAI11735.1 | |
DC335107.1 | 1 | 378 | ||
M64752.1 | 1 | 87 | AAA58613.1 | |
M81886.1 | 1 | 155 | AAA58395.1 | |
X58633.1 | 1 | 115 | CAA41491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1019051 | Chr.5:153171303 - 153678336 on Build GRCh38 | Gain | GRIA1 |
esv2657105 | Chr.5:153385677 - 153727589 on Build GRCh38 | Deletion | GRIA1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |