Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | NP_848611.2 | ||
AK092858.1 | BAC03991.1 | |||
AK092906.1 | BAC04000.1 | |||
AK292004.1 | ||||
AK300915.1 | ||||
DA945486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | AGRP NRN1L DPEP2 PARD6A GFOD2 NOL3 DPEP3 ELMO3 SLC9A5 ATP6V0D1 LOC107984813 LRRC36 HSF4 TSNAXIP1 LOC100505942 TMEM208 NUTF2 MIR328 CENPT FBXL8 E2F4 ACD CTCF HSD11B2 C16orf86 FHOD1 LRRC29 TPPP3 EDC4 LCAT TRADD CARMIL2 THAP11 PLEKHG4 DUS2 PSKH1 KIAA0895L FAM65A PSMB10 DDX28 LOC105369155 ZDHHC1 KCTD19 CTRL LOC100131303 RANBP10 EXOC3L1 ENKD1 SLC12A4 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | KIAA0895L FHOD1 LOC105369155 C16orf70 LRRC29 HSF4 TRADD PLEKHG4 NOL3 TMEM208 ELMO3 B3GNT9 MIR328 FBXL8 SLC9A5 EXOC3L1 E2F4 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | KIAA0895L FHOD1 LOC105369155 C16orf70 LRRC29 HSF4 TRADD NOL3 TMEM208 ELMO3 B3GNT9 MIR328 FBXL8 EXOC3L1 E2F4 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | KIAA0895L NOL3 ELMO3 LOC105369155 MIR328 FBXL8 LRRC29 EXOC3L1 HSF4 E2F4 |
nsv471092 | Chr.16:67174030 - 67518392 on Build GRCh38 | Gain | KIAA0895L HSD11B2 AGRP FHOD1 LOC105369155 ZDHHC1 LRRC36 LRRC29 TPPP3 KCTD19 LOC100505942 PLEKHG4 NOL3 TMEM208 ELMO3 MIR328 SLC9A5 EXOC3L1 E2F4 ATP6V0D1 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | AGRP NRN1L DPEP2 PARD6A GFOD2 NOL3 DPEP3 ELMO3 SLC9A5 ATP6V0D1 LOC107984813 C16orf70 LRRC36 HSF4 TSNAXIP1 LOC100505942 TMEM208 NUTF2 MIR328 CENPT FBXL8 E2F4 ACD CTCF HSD11B2 C16orf86 FHOD1 LRRC29 TPPP3 EDC4 LCAT TRADD CARMIL2 THAP11 PLEKHG4 DUS2 PSKH1 B3GNT9 KIAA0895L FAM65A PSMB10 DDX28 LOC105369155 ZDHHC1 KCTD19 CTRL LOC100131303 RANBP10 EXOC3L1 ENKD1 SLC12A4 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | KIAA0895L PLEKHG4 FHOD1 TMEM208 ELMO3 LOC105369155 MIR328 LRRC29 SLC9A5 EXOC3L1 E2F4 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |