Assay Details
Target Gene Details
Entrez Gene ID: | 3091 |
Gene Name: | hypoxia inducible factor 1 alpha subunit |
Gene Aliases: |
HIF-1-alpha, HIF-1A, HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PASD8, bHLHe78 |
Location: |
Chr.14:61695401-61748259 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HIF1A | NM_001243084.1 | 15 | 2727 | NP_001230013.1 |
NM_001530.3 | 15 | 2830 | NP_001521.1 | |
NM_181054.2 | 14 | 2703 | NP_851397.1 | |
AB073325.1 | 14 | 2327 | BAB70608.1 | |
AB500182.1 | 15 | 2451 | ||
AF207601.1 | 15 | 2436 | AAF20139.1 | |
AF207602.1 | 15 | 2433 | AAF20140.1 | |
AF304431.1 | 15 | 2426 | AAG43026.1 | |
AK296876.1 | ||||
AK299551.1 | 16 | 2532 | ||
AK304315.1 | 15 | 2539 | ||
AK304436.1 | 14 | 2279 | ||
AK312397.1 | 15 | 2659 | ||
BC012527.2 | 15 | 2680 | AAH12527.1 | |
BT009776.1 | 15 | 2426 | AAP88778.1 | |
BX648795.1 | 15 | 2694 | ||
CN264295.1 | 2 | 117 | ||
CN264320.1 | 4 | 561 | ||
FJ790247.1 | 15 | 2728 | ||
U22431.1 | 15 | 2454 | AAC50152.1 | |
U29165.1 | 15 | 2690 | AAC51210.1 | |
X72726.1 |
Target Gene Details
Entrez Gene ID: | 100750247 |
Gene Name: | HIF1A antisense RNA 2 |
Gene Aliases: |
3'aHIF-1A, aHIF |
Location: |
Chr.14:61747039-61749089 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HIF1A-AS2 | NR_045406.1 | |||
BF923330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1300 | Chr.14:61737786 - 61782490 on Build GRCh38 | Deletion | HIF1A SNAPC1 HIF1A-AS2 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |